Gene Transcripts

Gene Symbol
SNTA1
Entrez Gene ID
6640
Gene Name
syntrophin alpha 1
Gene Aliases
LQT12, SNT1, TACIP1, dJ1187J4.5
Chromosome Location
Chr. 20: 33407957 - 33443892 on Build GRCh38
UniGene ID
Hs.31121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003098.2 NP_003089.1 7 1545
XM_005260517.1 XP_005260574.1 7 1506
XM_011529007.1 XP_011527309.1 7 1538
XM_011529008.1 XP_011527310.1 7 1538
GenBank mRNA AK095942.1 - 7 1105
AK291994.1 - 7 1416
AK301800.1 - 7 1051
BC026215.2 - 7 1385 4413197
S81737.1 - 7 1405
U40571.1 - 7 1310

View Full Product Details