Gene Transcripts
- Gene Symbol
- Rps21
- Entrez Gene ID
- 81775
- Gene Name
- ribosomal protein S21
- Gene Aliases
- Chromosome Location
- Chr. 3: 175629087 - 175630205 on Build Rnor_6.0
- UniGene ID
- Rn.4153
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031111.1 |
NP_112373.1
|
4, 5 |
233 |
|
XM_006235823.1 |
XP_006235885.1
|
4, 5 |
322 |
|
GenBank mRNA |
BC058464.1 |
- |
4, 5 |
238 |
|
FQ210654.1 |
- |
4, 5 |
236 |
|
FQ210736.1 |
- |
4, 5 |
236 |
|
FQ210823.1 |
- |
4, 5 |
234 |
|
FQ210868.1 |
- |
4, 5 |
235 |
|
FQ212305.1 |
- |
4, 5 |
235 |
|
FQ212673.1 |
- |
4, 5 |
228 |
|
FQ217300.1 |
- |
4, 5 |
234 |
|
FQ221031.1 |
- |
4, 5 |
236 |
|
FQ221222.1 |
- |
4, 5 |
236 |
|
FQ221352.1 |
- |
4, 5 |
234 |
|
FQ221376.1 |
- |
4, 5 |
234 |
|
FQ221504.1 |
- |
4, 5 |
234 |
|
FQ221719.1 |
- |
4, 5 |
235 |
|
FQ221773.1 |
- |
4, 5 |
234 |
|
FQ221882.1 |
- |
4, 5 |
234 |
|
FQ221895.1 |
- |
4, 5 |
234 |
|
FQ222195.1 |
- |
4, 5 |
234 |
|
FQ222197.1 |
- |
4, 5 |
234 |
|
FQ222346.1 |
- |
4, 5 |
235 |
|
FQ222395.1 |
- |
4, 5 |
234 |
|
FQ222640.1 |
- |
4, 5 |
237 |
|
FQ222661.1 |
- |
4, 5 |
234 |
|
FQ222692.1 |
- |
4, 5 |
236 |
|
FQ222717.1 |
- |
4, 5 |
234 |
|
FQ222861.1 |
- |
4, 5 |
234 |
|
FQ223083.1 |
- |
4, 5 |
234 |
|
FQ223085.1 |
- |
4, 5 |
234 |
|
FQ223126.1 |
- |
4, 5 |
234 |
|
FQ223140.1 |
- |
4, 5 |
233 |
|
FQ223281.1 |
- |
4, 5 |
234 |
|
FQ223324.1 |
- |
4, 5 |
234 |
|
FQ223458.1 |
- |
4, 5 |
235 |
|
FQ223473.1 |
- |
4, 5 |
264 |
|
FQ224512.1 |
- |
4, 5 |
235 |
|
FQ224728.1 |
- |
4, 5 |
236 |
|
FQ228462.1 |
- |
4, 5 |
234 |
|
FQ228565.1 |
- |
4, 5 |
235 |
|
FQ228580.1 |
- |
4, 5 |
235 |
|
FQ228593.1 |
- |
4, 5 |
234 |
|
FQ228604.1 |
- |
4, 5 |
239 |
|
FQ229072.1 |
- |
4, 5 |
229 |
|
FQ229301.1 |
- |
4, 5 |
238 |
|
FQ229813.1 |
- |
4, 5 |
234 |
|
X79059.1 |
- |
4, 5 |
233 |
|
View Full Product Details