Gene Transcripts

Gene Symbol
Prl2b1
Entrez Gene ID
192224
Gene Name
Prolactin family 2, subfamily b, member 1
Gene Aliases
Prlpk
Chromosome Location
Chr. 17: 39542058 - 39549080 on Build Rnor_6.0
UniGene ID
Rn.19300
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138861.1 NP_620216.1 4 386
GenBank mRNA AB022882.1 - 4 383
AF234635.1 - 4 386

View Full Product Details