Gene Transcripts

Gene Symbol
VEGFB
Entrez Gene ID
7423
Gene Name
vascular endothelial growth factor B
Gene Aliases
VEGFL, VRF
Chromosome Location
Chr. 11: 64234584 - 64239264 on Build GRCh38
UniGene ID
Hs.732095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243733.1 NP_001230662.1 7 1099
NM_003377.4 NP_003368.1 7 1200
GenBank mRNA BC008818.2 - 7 990
BG031449.1 - 2 424
BM694361.1 - 1 41
U43368.1 - 7 952

View Full Product Details