Gene Transcripts

Gene Symbol
Mpdu1
Entrez Gene ID
303244
Gene Name
mannose-P-dolichol utilization defect 1
Gene Aliases
Chromosome Location
Chr. 10: 56261637 - 56267213 on Build Rnor_6.0
UniGene ID
Rn.17468
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107011.1 NP_001100481.1 1 90
GenBank mRNA BC087071.1 - 1 236
BC107940.1 - 1 49
FQ213141.1 - 1 66
FQ220130.1 - 1 54

View Full Product Details