Gene Transcripts

Gene Symbol
Srsf4
Entrez Gene ID
362612
Gene Name
serine/arginine-rich splicing factor 4
Gene Aliases
RGD1561347, Sfrs4
Chromosome Location
Chr. 5: 150032999 - 150060769 on Build Rnor_6.0
UniGene ID
Rn.63428
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108685.2 NP_001102155.1 4 563
GenBank mRNA BC160850.1 - 1 36

View Full Product Details