Gene Transcripts

Gene Symbol
SPHK1
Entrez Gene ID
8877
Gene Name
sphingosine kinase 1
Gene Aliases
SPHK
Chromosome Location
Chr. 17: 76382977 - 76387860 on Build GRCh38
UniGene ID
Hs.68061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142601.1 NP_001136073.1 3 546
NM_001142602.1 NP_001136074.1 2 591
NM_021972.3 NP_068807.2 3 588
NM_182965.2 NP_892010.2 3 909
XM_005257766.2 XP_005257823.1 3 901
GenBank mRNA AF200328.1 - 2 172
AF238083.1 - 3 490
AF266756.1 - 2 423
AJ245504.1 - 1 145
AK022402.1 - 3 546
AK023393.1 - 2 503
AK095578.1 - 2 1231
AK122781.1 - 5 963
AK225250.1 - 3 509
AK225258.1 - 3 509
AK292294.1 - 3 633
AK292614.1 - 2 507
BC008040.2 - 3 483 3831657
BC009419.1 - 3 576 3506924
BC014439.1 - 3 846 4871343
BC030553.1 - 3 868 5213270
DC384776.1 - 2 519

View Full Product Details