Gene Transcripts

Gene Symbol
MPDU1
Entrez Gene ID
9526
Gene Name
mannose-P-dolichol utilization defect 1
Gene Aliases
CDGIF, HBEBP2BPA, Lec35, My008, PP3958, PQLC5, SL15
Chromosome Location
Chr. 17: 7583647 - 7588212 on Build GRCh38
UniGene ID
Hs.246381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004870.3 NP_004861.2 3 493
NR_024603.1 3 488
XM_006721597.2 XP_006721660.1 3 292
XM_006721598.2 XP_006721661.1 3 292
XM_006721599.2 XP_006721662.1 3 292
XM_011524081.2 XP_011522383.1 3 288
GenBank mRNA AF038961.1 - 3 293
AF059752.1 - 3 333
AF258568.1 - 1239
AK055923.1 - 3 287
AK075299.1 - 3 292
AK225401.1 - 3 292
AK225432.1 - 3 288
AK297001.1 - 3 288
AK297691.1 - 3 288
AK300083.1 - 3 288
AK300140.1 - 3 279
AK314527.1 - 3 288
BC001898.1 - 3 304 3536216
BP325875.1 - 3 494
DC421138.1 - 3 494
DQ499597.1 - 3 277
KU178478.1 - 3 277
KU178479.1 - 3 277

View Full Product Details