Gene Transcripts

Gene Symbol
Smarcc1
Entrez Gene ID
301020
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1
Gene Aliases
Chromosome Location
Chr. 8: 118206689 - 118326264 on Build Rnor_6.0
UniGene ID
Rn.94211
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106861.1 NP_001100331.1 2 340
XM_006243770.2 XP_006243832.1 2 354
XM_006243771.2 XP_006243833.1 2 354
XM_006243772.2 XP_006243834.1 2 354

View Full Product Details