Gene Transcripts

Gene Symbol
Hus1b
Entrez Gene ID
210554
Gene Name
Hus1 homolog b (S. pombe)
Gene Aliases
Chromosome Location
Chr. 13: 30946576 - 30947761 on Build GRCm38
UniGene ID
Mm.450571
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153072.2 NP_694712.1 1 308
GenBank mRNA AF508546.1 - 1 221
AK133107.1 - 1 309
BC125607.1 - 1 301 40130122
BC125609.1 - 1 301 40130124

View Full Product Details