Gene Transcripts
- Gene Symbol
- Def8
- Entrez Gene ID
- 23854
- Gene Name
- differentially expressed in FDCP 8
- Gene Aliases
- AI449518, D8Ertd713e, DEF-8
- Chromosome Location
- Chr. 8: 123442956 - 123463899 on Build GRCm38
- UniGene ID
- Mm.275127
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253783.1 |
NP_001240712.1
|
7, 8 |
720 |
|
NM_001253784.1 |
NP_001240713.1
|
6, 7 |
698 |
|
NM_001281803.1 |
NP_001268732.1
|
5, 6 |
702 |
|
NM_054046.5 |
NP_473387.1
|
5, 6 |
611 |
|
GenBank mRNA |
AK028726.1 |
- |
6, 7 |
670 |
|
AK034028.1 |
- |
|
577 |
|
AK050405.1 |
- |
6, 7 |
669 |
|
AK081295.1 |
- |
5, 6 |
601 |
|
AK086174.1 |
- |
5, 6 |
572 |
|
AK087778.1 |
- |
5, 6 |
582 |
|
AK129019.1 |
- |
5, 6 |
653 |
|
AK133979.1 |
- |
5, 6 |
585 |
|
AK150334.1 |
- |
5, 6 |
547 |
|
AK150994.1 |
- |
5, 6 |
547 |
|
AK164147.1 |
- |
5, 6 |
583 |
|
BC003306.1 |
- |
7, 8 |
671 |
3154712 |
BY713285.1 |
- |
5, 6 |
612 |
|
CD545768.1 |
- |
1, 2 |
97 |
|
CX562217.1 |
- |
5, 6 |
664 |
|
View Full Product Details