Gene Transcripts

Gene Symbol
Cyp4f39
Entrez Gene ID
320997
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 39
Gene Aliases
4732474A20Rik
Chromosome Location
Chr. 17: 32452695 - 32493320 on Build GRCm38
UniGene ID
Mm.130900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177307.3 NP_796281.1 10, 11 1325
GenBank mRNA AK028950.1 - 10, 11 1326
AK037192.1 - 10, 11 1326
BC145755.1 - 10, 11 1322 40131058

View Full Product Details