Gene Transcripts

Gene Symbol
POLR3A
Entrez Gene ID
11128
Gene Name
RNA polymerase III subunit A
Gene Aliases
ADDH, HLD7, RPC1, RPC155, hRPC155
Chromosome Location
Chr. 10: 77975149 - 78029540 on Build GRCh38
UniGene ID
Hs.436896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007055.3 NP_008986.2 10 1473
GenBank mRNA AF021351.1 - 11 1340
AK026023.1 - 1 47
AK300329.1 - 10 1453
AY091460.1 - 3 248
BC041089.1 - 10 1434 3915501

View Full Product Details