Gene Transcripts

Gene Symbol
Klrh1
Entrez Gene ID
246043
Gene Name
killer cell lectin-like receptor subfamily H, member 1
Gene Aliases
Chromosome Location
Chr. 4: 163597634 - 163611383 on Build Rnor_6.0
UniGene ID
Rn.83604
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139187.2 NP_631926.1 3 227
GenBank mRNA AF416564.2 - 3 227

View Full Product Details