Gene Transcripts

Gene Symbol
OVOL3
Entrez Gene ID
728361
Gene Name
ovo like zinc finger 3
Gene Aliases
HOVO3
Chromosome Location
Chr. 19: 36111180 - 36113711 on Build GRCh38
UniGene ID
Hs.143920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017027190.1 XP_016882679.1 1 163

View Full Product Details