Gene Transcripts

Gene Symbol
KNCN
Entrez Gene ID
148930
Gene Name
kinocilin
Gene Aliases
Kino, L5
Chromosome Location
Chr. 1: 46545638 - 46551657 on Build GRCh38
UniGene ID
Hs.350764
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097611.1 NP_001091080.1 1 123
NM_001322255.1 NP_001309184.1 1 565
GenBank mRNA DA208279.1 - 1 39

View Full Product Details