Gene Transcripts

Gene Symbol
Cnfn
Entrez Gene ID
72383
Gene Name
cornifelin
Gene Aliases
2210418J09Rik
Chromosome Location
Chr. 7: 25367616 - 25369724 on Build GRCm38
UniGene ID
Mm.34382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081375.1 NP_001074844.1 4 391
NM_028219.1 NP_082495.1 5 584
GenBank mRNA AK008979.1 - 4 392
BC059093.1 - 5 563 30293557

View Full Product Details