Gene Transcripts

Gene Symbol
SAR1B
Entrez Gene ID
51128
Gene Name
secretion associated Ras related GTPase 1B
Gene Aliases
ANDD, CMRD, GTBPB, SARA2
Chromosome Location
Chr. 5: 134601149 - 134632843 on Build GRCh38
UniGene ID
Hs.432984
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033503.2 NP_001028675.1 4 346
NM_016103.3 NP_057187.1 3 227
XM_017009521.1 XP_016865010.1 3 234
GenBank mRNA AF087850.1 - 3 264
AF092130.1 - 3 243
AK223452.1 - 3 212
AL710660.1 - 4 328
BC002847.2 - 4 277 3636540
BC093034.1 - 3 227 30528518
CD048852.1 - 4 354

View Full Product Details