Gene Transcripts

Gene Symbol
DMRT3
Entrez Gene ID
58524
Gene Name
doublesex and mab-3 related transcription factor 3
Gene Aliases
DMRTA3
Chromosome Location
Chr. 9: 976968 - 991732 on Build GRCh38
UniGene ID
Hs.189174
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021240.3 NP_067063.1 2 939
GenBank mRNA AF193873.1 - 2 520
BC113584.1 - 2 939 8322636
BC117245.1 - 2 939 40125796

View Full Product Details