Gene Transcripts

Gene Symbol
POPDC2
Entrez Gene ID
64091
Gene Name
popeye domain containing 2
Gene Aliases
POP2
Chromosome Location
Chr. 3: 119642052 - 119660590 on Build GRCh38
UniGene ID
Hs.534383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308333.1 NP_001295262.1 5 1287
NM_022135.3 NP_071418.2 4 1204
XM_005247698.3 XP_005247755.1 4 1345
GenBank mRNA AF204173.1 - 4 1193
AK124602.1 - 1 4438
AK226022.1 - 6 1493
AK314819.1 - 5 1288
AK377174.1 - 7 684
AK377176.1 - 7 962
AK377177.1 - 5 966
AK377178.1 - 7 1020
AK377179.1 - 6 1052
AK377180.1 - 8 1093
AK377181.1 - 7 1101
AK377182.1 - 7 1161
AK377183.1 - 8 1222
AK377184.1 - 8 1234
BC026911.2 - 6 834
BC044929.1 - 4 1204 5191230

View Full Product Details