Gene Transcripts

Gene Symbol
THSD7B
Entrez Gene ID
80731
Gene Name
thrombospondin type 1 domain containing 7B
Gene Aliases
Chromosome Location
Chr. 2: 136765562 - 137677712 on Build GRCh38
UniGene ID
Hs.68533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316349.1 NP_001303278.1 21 3981
XM_017005049.1 XP_016860538.1 15 2458
GenBank mRNA AB051466.1 - 20 3611

View Full Product Details