Gene Transcripts

Gene Symbol
OR2G2
Entrez Gene ID
81470
Gene Name
olfactory receptor family 2 subfamily G member 2
Gene Aliases
OR1-32
Chromosome Location
Chr. 1: 247588360 - 247589313 on Build GRCh38
UniGene ID
Hs.690208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001915.1 NP_001001915.1 1 684

View Full Product Details