Gene Transcripts

Gene Symbol
PIK3IP1
Entrez Gene ID
113791
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
HGFL, hHGFL(S)
Chromosome Location
Chr. 22: 31281593 - 31292534 on Build GRCh38
UniGene ID
Hs.26670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135911.1 NP_001129383.1 1 233
NM_052880.4 NP_443112.2 1 233
GenBank mRNA AF528079.1 - 1 178
AF528080.1 - 1 178
AK074688.1 - 1 179
AK226089.1 - 1 113
AK299397.1 - 1 187
AK300990.1 - 1 157
BC011049.2 - 1 178 4180134
BQ718653.1 - 1 211
CR456340.1 - 1 73
DC412134.1 - 1 233
GU135609.1 - 1 94

View Full Product Details