Gene Transcripts

Gene Symbol
OR5P2
Entrez Gene ID
120065
Gene Name
olfactory receptor family 5 subfamily P member 2
Gene Aliases
JCG3, JCG4
Chromosome Location
Chr. 11: 7795974 - 7796942 on Build GRCh38
UniGene ID
Hs.351824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153444.1 NP_703145.1 1 103
GenBank mRNA AF173006.1 - 1 103
BC101710.1 - 1 134 8069216
BC113557.1 - 1 134 8322609

View Full Product Details