Gene Transcripts
- Gene Symbol
- STK32C
- Entrez Gene ID
- 282974
- Gene Name
- serine/threonine kinase 32C
- Gene Aliases
- PKE, YANK3
- Chromosome Location
- Chr. 10: 132207482 - 132332559 on Build GRCh38
- UniGene ID
- Hs.469002
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318878.1 |
NP_001305807.1
|
10, 11 |
1437 |
|
NM_001318879.1 |
NP_001305808.1
|
10, 11 |
1298 |
|
NM_173575.3 |
NP_775846.2
|
10, 11 |
1399 |
|
XM_011539688.1 |
XP_011537990.1
|
10, 11 |
1470 |
|
XM_011539690.2 |
XP_011537992.1
|
10, 11 |
1385 |
|
XM_011539693.2 |
XP_011537995.1
|
10, 11 |
1403 |
|
XM_011539694.1 |
XP_011537996.1
|
10, 11 |
1137 |
|
XM_011539695.1 |
XP_011537997.1
|
10, 11 |
1338 |
|
XM_011539696.1 |
XP_011537998.1
|
10, 11 |
1145 |
|
GenBank mRNA |
AK098366.1 |
- |
8, 9 |
1426 |
|
AK302102.1 |
- |
10, 11 |
1384 |
|
AY098866.1 |
- |
10, 11 |
1399 |
|
BC045760.1 |
- |
10, 11 |
1288 |
4831007 |
View Full Product Details