Gene Transcripts

Gene Symbol
C2orf61
Entrez Gene ID
285051
Gene Name
chromosome 2 open reading frame 61
Gene Aliases
Chromosome Location
Chr. 2: 47086991 - 47155378 on Build GRCh38
UniGene ID
Hs.531575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163561.1 NP_001157033.1 3 398
NM_173649.2 NP_775920.1 3 398
GenBank mRNA AK097491.1 - 3 307
DA313162.1 - 3 398

View Full Product Details