Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001029955.3 | NP_001025126.2 | 1 | 795 | |
GenBank mRNA | AK122914.1 | - | 1 | 713 | |
BC035027.1 | - | 1 | 668 | 4828469 | |
BC052943.1 | - | 1 | 735 | 5268527 | |
BC104224.1 | - | 1 | 717 | 40032133 | |
BC104225.1 | - | 1 | 717 | 40032134 |