Gene Transcripts
- Gene Symbol
- LOC647115
- Entrez Gene ID
- 647115
- Gene Name
- translation initiation factor IF-2
- Gene Aliases
- Chromosome Location
- Chr. 2: 63841107 - 63842137 on Build GRCh38
- UniGene ID
- Hs.516217
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094167.1 |
- |
1 |
1838 |
|
- Gene Symbol
- WDPCP
- Entrez Gene ID
- 51057
- Gene Name
- WD repeat containing planar cell polarity effector
- Gene Aliases
- BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
- Chromosome Location
- Chr. 2: 63120755 - 63840825 on Build GRCh38
- UniGene ID
- Hs.414952
- Species
- Homo sapiens
View Full Product Details