Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004963.3 | NP_004954.2 | 4 | 711 | |
XM_011520631.2 | XP_011518933.1 | 4 | 609 | ||
GenBank mRNA | AK310034.1 | - | 4 | 701 | |
BC136544.1 | - | 4 | 620 | 9020534 | |
BC136545.1 | - | 4 | 620 | 9020535 | |
M73489.1 | - | 4 | 622 | ||
S57551.1 | - | 4 | 691 |