Gene Transcripts
- Gene Symbol
- Pax4
- Entrez Gene ID
- 18506
- Gene Name
- paired box 4
- Gene Aliases
- Pax-4
- Chromosome Location
- Chr. 6: 28442334 - 28449340 on Build GRCm38
- UniGene ID
- Mm.8026
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159925.1 |
NP_001153397.1
|
7, 8 |
917 |
|
NM_001159926.1 |
NP_001153398.1
|
7, 8 |
917 |
|
NM_011038.2 |
NP_035168.1
|
7, 8 |
917 |
|
GenBank mRNA |
AB008912.1 |
- |
7, 8 |
892 |
|
AB010557.1 |
- |
7, 8 |
860 |
|
AB010558.1 |
- |
7, 8 |
875 |
|
AF031150.1 |
- |
7, 8 |
950 |
|
AF104231.1 |
- |
6, 7 |
672 |
|
AF145233.1 |
- |
7, 8 |
864 |
|
BC131906.1 |
- |
7, 8 |
828 |
40130183 |
BC131908.1 |
- |
7, 8 |
826 |
40130185 |
BC145168.1 |
- |
7, 8 |
828 |
9053719 |
BC145169.1 |
- |
7, 8 |
828 |
9053720 |
Y09584.1 |
- |
5, 6 |
556 |
|
View Full Product Details