Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011172.2 | NP_035302.2 | 11, 12 | 1460 | |
GenBank mRNA | AF120279.1 | - | 11, 12 | 1417 | |
AK144068.1 | - | 11, 12 | 1430 | ||
BC008548.1 | - | 11, 12 | 1284 | ||
BC125327.1 | - | 11, 12 | 1263 | 40129842 | |
U80020.1 | - | 11, 12 | 1409 |