Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083937.1 | NP_001077406.1 | 2 | 209 | |
NM_078484.2 | NP_511039.2 | 2 | 209 | ||
GenBank mRNA | AB027147.1 | - | 2 | 128 | |
AF229634.1 | - | 2 | 209 | ||
AK019529.1 | - | 1 | 1729 | ||
AK146830.1 | - | 2 | 125 | ||
AK155755.1 | - | 2 | 121 | ||
BC037701.1 | - | 2 | 146 | 4187325 |