Gene Transcripts
- Gene Symbol
- Reg3d
- Entrez Gene ID
- 30053
- Gene Name
- regenerating islet-derived 3 delta
- Gene Aliases
- INGAP, Ingaprp
- Chromosome Location
- Chr. 6: 78375874 - 78378865 on Build GRCm38
- UniGene ID
- Mm.33691
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB028625.1 |
- |
6 |
551 |
|
View Full Product Details