Gene Transcripts
- Gene Symbol
- Rps21
- Entrez Gene ID
- 66481
- Gene Name
- ribosomal protein S21
- Gene Aliases
- 1810049N11Rik, 2410030A14Rik
- Chromosome Location
- Chr. 2: 180257373 - 180258444 on Build GRCm38
- UniGene ID
- Mm.289669
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025587.2 |
NP_079863.1
|
3, 4 |
201 |
|
GenBank mRNA |
AA049954.1 |
- |
2, 3 |
128 |
|
AK007846.1 |
- |
3, 4 |
164 |
|
AK007850.1 |
- |
3, 4 |
156 |
|
AK010610.1 |
- |
3, 4 |
202 |
|
AK010637.1 |
- |
3, 4 |
169 |
|
AK012488.1 |
- |
3, 4 |
164 |
|
BC027563.1 |
- |
3, 4 |
149 |
3468174 |
BC086912.1 |
- |
3, 4 |
166 |
6758965 |
BU534365.1 |
- |
3, 4 |
204 |
|
View Full Product Details