Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005417.2 | NP_001005417.1 | 7 | 1222 | |
NM_003780.4 | NP_003771.1 | 7 | 1422 | ||
NM_030587.2 | NP_085076.2 | 7 | 1223 | ||
XM_017002716.1 | XP_016858205.1 | 7 | 1180 | ||
XM_017002717.1 | XP_016858206.1 | 7 | 1383 | ||
GenBank mRNA | AB024434.1 | - | 6 | 1068 | |
AF038660.1 | - | 6 | 1112 | ||
AK074533.1 | - | 5 | 720 | ||
AK095873.1 | - | 7 | 1850 | ||
AK293418.1 | - | 7 | 1172 | ||
AL137647.1 | - | 5 | 617 | ||
BC096821.1 | - | 7 | 1190 | 30558762 | |
Y12510.1 | - | 6 | 1060 |