Gene Transcripts

Gene Symbol
MUC7
Entrez Gene ID
4589
Gene Name
mucin 7, secreted
Gene Aliases
MG2
Chromosome Location
Chr. 4: 70430492 - 70482997 on Build GRCh38
UniGene ID
Hs.631946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145006.1 NP_001138478.1 4 739
NM_001145007.1 NP_001138479.1 4 678
NM_152291.2 NP_689504.2 3 641
GenBank mRNA AK093828.1 - 4 425
AK312855.1 - 3 559
BC025688.1 - 3 539 5205526
BX537739.1 - 1 714
BX647190.1 - 4 443
BX648903.1 - 1 366
BX648904.1 - 1 292
L13283.1 - 3 547

View Full Product Details