Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174989.4 | NP_778154.1 | 2 | 2240 | |
GenBank mRNA | AB091053.1 | - | 1 | 2107 | |
AK054219.1 | - | 2 | 2200 | ||
AK083836.1 | - | 2 | 2218 | ||
AK086237.1 | - | 2 | 2218 | ||
AK089679.1 | - | 2 | 2218 | ||
AK143766.1 | - | 2 | 2232 | ||
AK150150.1 | - | 2 | 2234 | ||
AK155245.1 | - | 2 | 2240 | ||
AK155906.1 | - | 2 | 2232 | ||
AK171578.1 | - | 2 | 2232 | ||
BC033406.1 | - | 2 | 959 | ||
BC037048.1 | - | 2 | 2240 | ||
BC062191.1 | - | 2 | 885 | ||
BC094338.1 | - | 1 | 2176 | 30606722 |