Gene Transcripts

Gene Symbol
ZNF215
Entrez Gene ID
7762
Gene Name
zinc finger protein 215
Gene Aliases
BAZ-2, BAZ2, ZKSCAN11, ZSCAN43
Chromosome Location
Chr. 11: 6926423 - 6958047 on Build GRCh38
UniGene ID
Hs.523457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013250.2 NP_037382.2 7 1360
XM_006718311.2 XP_006718374.1 7 1400
GenBank mRNA AF056618.1 - 7 1360

View Full Product Details