Gene Transcripts

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
Chromosome Location
Chr. 3: 4493348 - 4847840 on Build GRCh38
UniGene ID
Hs.567295
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099952.2 NP_001093422.2 14 1560
NM_001168272.1 NP_001161744.1 13 1515
NM_002222.5 NP_002213.5 13 1515
XM_005265109.2 XP_005265166.1 14 1560
XM_005265110.2 XP_005265167.1 14 1560
XM_006713131.2 XP_006713194.1 14 1560
XM_011533681.1 XP_011531983.1 14 1560
XM_011533682.2 XP_011531984.1 14 1486
XM_011533683.2 XP_011531985.1 14 1560
XM_011533684.1 XP_011531986.1 14 1560
XM_011533685.1 XP_011531987.1 14 1560
XM_011533686.1 XP_011531988.1 14 1560
XM_011533687.1 XP_011531989.1 13 1515
XM_011533688.1 XP_011531990.1 13 1515
XM_011533690.1 XP_011531992.1 14 1560
XM_011533691.1 XP_011531993.1 14 1560
XM_011533692.2 XP_011531994.1 14 1560
XM_017006357.1 XP_016861846.1 14 1560
XM_017006358.1 XP_016861847.1 14 1560
GenBank mRNA AK293752.1 - 8 746
D26070.1 - 13 1421
L38019.2 - 14 1457
U23850.1 - 11 1165

View Full Product Details