Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127219.1 | NP_001120691.1 | 9 | 926 | |
NM_032592.3 | NP_115981.1 | 9 | 1222 | ||
XM_005253170.3 | XP_005253227.2 | 9 | 1365 | ||
XM_005253172.2 | XP_005253229.1 | 6 | 453 | ||
XM_006718348.2 | XP_006718411.1 | 9 | 674 | ||
XM_011520406.2 | XP_011518708.1 | 9 | 1368 | ||
XM_011520407.1 | XP_011518709.1 | 9 | 718 | ||
XM_011520410.2 | XP_011518712.1 | 9 | 1619 | ||
GenBank mRNA | AK057649.1 | - | 9 | 1216 | |
AL833533.1 | - | 2 | 1473 | ||
AY026508.1 | - | 9 | 1177 | ||
BC020197.1 | - | 9 | 1196 | 3632065 | |
KU178811.1 | - | 8 | 782 | ||
KU178812.1 | - | 8 | 782 | ||
KU178813.1 | - | 7 | 722 |