Gene Transcripts
- Gene Symbol
- RAB23
- Entrez Gene ID
- 51715
- Gene Name
- RAB23, member RAS oncogene family
- Gene Aliases
- HSPC137
- Chromosome Location
- Chr. 6: 57186992 - 57222314 on Build GRCh38
- UniGene ID
- Hs.555016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278666.1 |
NP_001265595.1
|
2, 3 |
337 |
|
NM_001278667.1 |
NP_001265596.1
|
2, 3 |
509 |
|
NM_001278668.1 |
NP_001265597.1
|
2, 3 |
336 |
|
NM_016277.4 |
NP_057361.3
|
2, 3 |
806 |
|
NM_183227.2 |
NP_899050.1
|
2, 3 |
360 |
|
GenBank mRNA |
AB025427.1 |
- |
1, 2 |
152 |
|
AB034244.1 |
- |
2, 3 |
305 |
|
AF161486.1 |
- |
2, 3 |
302 |
|
AF498951.1 |
- |
1, 2 |
152 |
|
AK313796.1 |
- |
2, 3 |
357 |
|
AU280196.1 |
- |
2, 3 |
336 |
|
AY585189.1 |
- |
1, 2 |
152 |
|
BC015021.1 |
- |
2, 3 |
772 |
3910708 |
CR749371.1 |
- |
3, 4 |
540 |
|
DA021294.1 |
- |
2, 3 |
360 |
|
DA997359.1 |
- |
2, 3 |
505 |
|
View Full Product Details