Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr. 3: 14124940 - 14143680 on Build GRCh38
UniGene ID
Hs.517817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024334.2 NP_077310.1 5, 6 690
XM_017007176.1 XP_016862665.1 6, 7 812
GenBank mRNA AK027757.1 - 5, 6 616
AK027877.1 - 5, 6 554
AK074073.1 - 3, 4 226
AK075010.1 - 5, 6 541
AK225589.1 - 5, 6 613
AL136916.1 - 5, 6 584
AY358625.1 - 5, 6 453
BC003125.1 - 5, 6 585 3503636
BC008054.2 - 3, 4 180
BC011719.2 - 5, 6 585 4079766
KU178769.1 - 4, 5 436
KU178770.1 - 4, 5 436

View Full Product Details