Gene Transcripts

Gene Symbol
TTC30A
Entrez Gene ID
92104
Gene Name
tetratricopeptide repeat domain 30A
Gene Aliases
IFT70A
Chromosome Location
Chr. 2: 177614298 - 177618966 on Build GRCh38
UniGene ID
Hs.128384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152275.3 NP_689488.3 1 2887
GenBank mRNA AK125104.1 - 2 2591
AL049329.1 - 1 987
BC042848.1 - 1 2889 4831326

View Full Product Details