Gene Transcripts
- Gene Symbol
- ASXL1
- Entrez Gene ID
- 171023
- Gene Name
- additional sex combs like 1, transcriptional regulator
- Gene Aliases
- BOPS, MDS
- Chromosome Location
- Chr. 20: 32358062 - 32439319 on Build GRCh38
- UniGene ID
- Hs.374043
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015338.5 |
NP_056153.2
|
8, 9 |
1158 |
|
XM_006723727.3 |
XP_006723790.1
|
7, 8 |
1437 |
|
XM_006723728.3 |
XP_006723791.1
|
8, 9 |
767 |
|
XM_006723730.3 |
XP_006723793.1
|
8, 9 |
963 |
|
XM_006723732.2 |
XP_006723795.1
|
7, 8 |
786 |
|
XM_006723733.1 |
XP_006723796.1
|
1, 2 |
792 |
|
XM_011528648.2 |
XP_011526950.1
|
8, 9 |
1082 |
|
XM_011528652.2 |
XP_011526954.1
|
9, 10 |
2100 |
|
XM_017027704.1 |
XP_016883193.1
|
8, 9 |
2223 |
|
XM_017027705.1 |
XP_016883194.1
|
8, 9 |
1236 |
|
XM_017027706.1 |
XP_016883195.1
|
7, 8 |
1287 |
|
GenBank mRNA |
AB023195.2 |
- |
2, 3 |
208 |
|
AB084280.1 |
- |
7, 8 |
711 |
|
AJ438952.2 |
- |
8, 9 |
984 |
|
AK122923.1 |
- |
7, 8 |
786 |
|
AL117518.1 |
- |
1, 2 |
128 |
|
BC137278.1 |
- |
9, 10 |
921 |
9021275 |
BC137280.1 |
- |
9, 10 |
921 |
9021277 |
View Full Product Details