Gene Transcripts

Gene Symbol
OR12D3
Entrez Gene ID
81797
Gene Name
olfactory receptor family 12 subfamily D member 3
Gene Aliases
hs6M1-27
Chromosome Location
Chr. 6: 29373423 - 29375291 on Build GRCh38
UniGene ID
Hs.272280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030959.2 NP_112221.1 1 1067
GenBank mRNA AJ459858.1 - 1 1067
BC103892.1 - 1 1067 40005054

View Full Product Details