Gene Transcripts

Gene Symbol
SLA2
Entrez Gene ID
84174
Gene Name
Src like adaptor 2
Gene Aliases
C20orf156, MARS, SLAP-2, SLAP2
Chromosome Location
Chr. 20: 36612519 - 36646216 on Build GRCh38
UniGene ID
Hs.713578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032214.3 NP_115590.1 8 1687
NM_175077.2 NP_778252.1 8 1637
GenBank mRNA AK025645.1 - 8 1324
AK291513.1 - 8 1667
BC042041.1 - 8 1626 4429896

View Full Product Details