Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016946.4 | NP_058642.1 | 10 | 1440 | |
GenBank mRNA | AF154005.1 | - | 10 | 1112 | |
AF172398.2 | - | 10 | 1316 | ||
AF207907.1 | - | 10 | 1185 | ||
AK026665.1 | - | 10 | 1231 | ||
AK075152.1 | - | 10 | 1240 | ||
AK304412.1 | - | 9 | 1046 | ||
AL136649.1 | - | 10 | 1244 | ||
AY358896.1 | - | 10 | 1221 | ||
BC001533.2 | - | 10 | 1214 | 2990019 |