Gene Transcripts

Gene Symbol
RAD17
Entrez Gene ID
5884
Gene Name
RAD17 checkpoint clamp loader component
Gene Aliases
CCYC, HRAD17, R24L, RAD17SP, RAD24
Chromosome Location
Chr. 5: 69369297 - 69414801 on Build GRCh38
UniGene ID
Hs.16184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278622.1 NP_001265551.1 7 1119
NM_002873.1 NP_002864.1 6 1224
NM_133338.2 NP_579916.1 8 1120
NM_133339.2 NP_579917.1 5 677
NM_133340.2 NP_579918.1 5 710
NM_133341.2 NP_579919.1 5 636
NM_133342.2 NP_579920.1 7 1097
NM_133343.1 NP_579921.1 7 1062
NM_133344.2 NP_579922.1 6 879
XM_017009679.1 XP_016865168.1 7 939
XM_017009680.1 XP_016865169.1 6 993
XM_017009681.1 XP_016865170.1 6 851
GenBank mRNA AF017748.1 - 6 849
AF076838.1 - 6 1224
AF085736.2 - 5 677
AF098533.1 - 5 656
AF098534.1 - 5 582
AF112263.1 - 5 701
AF126424.1 - 8 1119
AJ001642.1 - 6 888
AJ004977.1 - 6 884
AK292487.1 - 7 1097
AL122068.1 - 7 1062
BC032304.1 - 5 690 5170876
BX537441.1 - 7 1119

View Full Product Details