Gene Transcripts

Gene Symbol
OR6A2
Entrez Gene ID
8590
Gene Name
olfactory receptor family 6 subfamily A member 2
Gene Aliases
I7, OR11-55, OR6A1, OR6A2P
Chromosome Location
Chr. 11: 6794525 - 6795908 on Build GRCh38
UniGene ID
Hs.652252
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003696.2 NP_003687.2 1 1022
GenBank mRNA BC101489.1 - 1 897
BC101491.1 - 1 897 8068997

View Full Product Details