Gene Transcripts

Gene Symbol
MRFAP1L1
Entrez Gene ID
114932
Gene Name
Morf4 family associated protein 1 like 1
Gene Aliases
PP784
Chromosome Location
Chr. 4: 6707701 - 6709879 on Build GRCh38
UniGene ID
Hs.593159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203462.2 NP_982287.1 2 1191
XM_005247932.1 XP_005247989.1 2 1182
GenBank mRNA AF155654.1 - 2 1187
AF258591.1 - 1 1726
BC008087.1 - 2 1138 3446352
BC066897.1 - 2 1191 4818881

View Full Product Details